Learn More
Bis(methyldiphenylphosphine)palladium(II)Dichloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Bis(methyldiphenylphosphine)palladium(II)Dichloride |
| CAS | 52611-08-2 |
| Molecular Formula | [C13H13P]2.Cl2Pd |
| Quantity | 500 mg |
| Synonym | Palladium, dichlorobis(methyldiphenylphosphine)-, Dichlorobis(methyldiphenylphosphine)palladium, Phosphine, methyldiphenyl-, palladium complex, Bis[methyldiphenyl)phosphine]palladium dichloride, Dichlorobis[methylene-bis(diphenylphosphine)]dipalladium(II), NSC 148042, Bis(methyldiphenylphosphine)palladium(II) dichloride |
| InChI Formula | InChI=1S/2C13H13P.2ClH.Pd/c2*1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13;;;/h2*2-11H,1H3;2*1H;/q;;;;+2/p-2 |
| SMILES | [Cl-][Pd+2]([Cl-])([P](C)(c1ccccc1)c2ccccc2)[P](C)(c3ccccc3)c4ccccc4 |
| IUPAC Name | dichloropalladium;methyl(diphenyl)phosphanium |
| Molecular Weight (g/mol) | 577.76 |
| Formula Weight | 575.9922 |