missing translation for 'onlineSavingsMsg'
Learn More

Bis(diethylamino)phosphorochloridate, 97%, Thermo Scientific™

Product Code. 11487074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487074 5 g 5g
11497074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487074 Supplier Thermo Scientific Alfa Aesar Supplier No. B21519.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794-24-7
Molecular Formula C8H20ClN2OP
Molecular Weight (g/mol) 226.68
MDL Number MFCD00137958
InChI Key HKKBJRUSFKTHSU-UHFFFAOYSA-N
Synonym bis diethylamino phosphoryl chloride, tetraethylphosphorodiamidic chloride, bis diethylamino chlorophosphine oxide, didiethylaminophosphinoyl chloride, n,n,n',n'-tetraethylphosphorodiamidic chloride, phosphorodiamidic chloride, tetraethyl, tetraethylphosphorodiamidic acid chloride, bis diethylamino phosphochloridate, bis diethylamino phosphinoyl chloride, phosphorodiamidicchloride, n,n,n',n'-tetraethyl
PubChem CID 74528
IUPAC Name N-[chloro(diethylamino)phosphoryl]-N-ethylethanamine
SMILES CCN(CC)P(Cl)(=O)N(CC)CC

Specifications

Density 1.07
Boiling Point 101°C (0.3 mmHg)
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 226.69
Percent Purity 97%
Chemical Name or Material Bis(diethylamino)phosphorochloridate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.