missing translation for 'onlineSavingsMsg'
Learn More

Bis(diethylamino)chlorophosphine, 94%

Product Code. p-7025510
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11460097

Brand: Thermo Scientific Alfa Aesar B21019.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 685-83-6 | C8H20ClN2P | 210.686 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685-83-6
Molecular Formula C8H20ClN2P
Molecular Weight (g/mol) 210.686
MDL Number MFCD00015493
InChI Key JVEHJSIFWIIFHM-UHFFFAOYSA-N
Synonym bis diethylamino chlorophosphine, bis diethylamino phosphorchloridite, n,n,n',n'-tetraethylphosphorodiamidous chloride, bdapcd, chloro diethylamino phosphanyl diethylamine, pubchem6475, phosphorodiamidous chloride, tetraethyl, chlorobis diethylamino phosphine, bisdiethylamino-chloro-phosphine, chloro-bis diethylamino-phosphine
PubChem CID 2733351
IUPAC Name N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine
SMILES CCN(CC)P(N(CC)CC)Cl

Specifications

Density 1.002
Boiling Point 55°C to 57°C (0.2 mmHg)
Flash Point 101°C (214°F)
Odor Acrid
Linear Formula [(C2H5)2N]2PCl
Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 210.69
Percent Purity 94%
Chemical Name or Material Bis(diethylamino)chlorophosphine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.