Learn More
Bis(diethylamino)chlorophosphine, 94%
Product Code. 11480097
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480097 100 g 100g
11460097 5 g 5g
11470097 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11480097 Supplier Thermo Scientific Alfa Aesar Supplier No. B21019.22
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 685-83-6 | C8H20ClN2P | 210.686 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 685-83-6
Molecular Formula C8H20ClN2P
Molecular Weight (g/mol) 210.686
MDL Number MFCD00015493
InChI Key JVEHJSIFWIIFHM-UHFFFAOYSA-N
Synonym bis diethylamino chlorophosphine, bis diethylamino phosphorchloridite, n,n,n',n'-tetraethylphosphorodiamidous chloride, bdapcd, chloro diethylamino phosphanyl diethylamine, pubchem6475, phosphorodiamidous chloride, tetraethyl, chlorobis diethylamino phosphine, bisdiethylamino-chloro-phosphine, chloro-bis diethylamino-phosphine
PubChem CID 2733351
IUPAC Name N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine
SMILES CCN(CC)P(N(CC)CC)Cl
Specifications
Density 1.002
Boiling Point 55°C to 57°C (0.2 mmHg)
Flash Point 101°C (214°F)
Odor Acrid
Linear Formula [(C2H5)2N]2PCl
Quantity 100 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 210.69
Percent Purity 94%
Chemical Name or Material Bis(diethylamino)chlorophosphine