Learn More
Bis(4-octylphenyl)amine, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(4-octylphenyl)amine |
| CAS | 101-67-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H43 N |
| Quantity | 250 mg |
| Synonym | Vanlube-81, p, p’-dioctyldiphenylamine |
| InChI Formula | InChI=1S/C28H43N/c1-3-5-7-9-11-13-15-25-17-21-27(22-18-25)29-28-23-19-26(20-24-28)16-14-12-10-8-6-4-2/h17-24,29H,3-16H2,1-2H3 |
| SMILES | CCCCCCCCc1ccc(Nc2ccc(CCCCCCCC)cc2)cc1 |
| IUPAC Name | 4-octyl-N-(4-octylphenyl)aniline |
| Molecular Weight (g/mol) | 393.65 |
| Show More |