Learn More
Bis(4-methoxyphenyl)phosphinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(4-methoxyphenyl)phosphinic Acid |
| CAS | 20434-05-3 |
| Molecular Formula | C14H15O4P |
| Quantity | 100 mg |
| Synonym | P,P-Bis(4-methoxyphenyl)phosphinic Acid |
| InChI Formula | InChI=1S/C14H15O4P/c1-17-11-3-7-13(8-4-11)19(15,16)14-9-5-12(18-2)6-10-14/h3-10H,1-2H3,(H,15,16) |
| SMILES | COC(C=C1)=CC=C1P(C2=CC=C(OC)C=C2)(O)=O |
| IUPAC Name | bis(4-methoxyphenyl)phosphinic acid |
| Molecular Weight (g/mol) | 278.24 |
| Formula Weight | 278.07 |