Learn More
Bis(2,5-iodomethyl)dioxane(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2,5-iodomethyl)dioxane (Mixture of Diastereomers) |
| CAS | 101084-46-2 |
| Molecular Formula | C6H10I2O2 |
| Quantity | 500 mg |
| Synonym | 2,5-Bis(iodomethyl)-p-dioxane |
| InChI Formula | InChI=1S/C6H10I2O2/c7-1-5-3-10-6(2-8)4-9-5/h5-6H,1-4H2 |
| SMILES | ICC1COC(CI)CO1 |
| IUPAC Name | 2,5-bis(iodomethyl)-1,4-dioxane |
| Molecular Weight (g/mol) | 367.95 |
| Formula Weight | 367.877 |