missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0) is used to prepare palladium(0) catalysts for highly selective cross-coupling reactions especially carbohalogenation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink to White |
Quantity | 1 g |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 1527.86 |
Physical Form | Crystalline |
Chemical Name or Material | Bis[1,2,3,4,5-pentaphenyl-1'- (di-tert-butylphosphino)ferrocene]palladium (0) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'