Learn More
Birinapant, MedChemExpress
Description
Birinapant (TL32711) induces the autoubiquitylation and proteasomal degradation of cIAP1 and cIAP2 in intact cells, which results in formation of a RIPK1: caspase-8 complex, caspase-8 activation, and induction of tumor cell death. Birinapant (TL32711) targets TRAF2-associated cIAPs and abrogates TNF-induced NF-κB activation.
Specifications
Specifications
| Chemical Name or Material | Birinapant |
| CAS | 1260251-31-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C42H56F2N8O6 |
| Quantity | 5 mg |
| Synonym | TL32711 |
| Solubility Information | DMSO : 125 mg/mL (154.91 mM; Need ultrasonic) |
| SMILES | FC1=CC2=C(C=C1)C(C[C@H]3N(C([C@@H](NC([C@@H](NC)C)=O)CC)=O)C[C@@H](O)C3)=C(C(N4)=C(C[C@H]5N(C([C@@H](NC([C@@H](NC)C)=O)CC)=O)C[C@@H](O)C5)C6=C4C=C(F)C=C6)N2 |
| Molecular Weight (g/mol) | 806.94 |
| Show More |