missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BirA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 326.00 €
Specifications
| Antigen | BirA |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621849
|
Novus Biologicals
NBP2-99832-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648657
|
Novus Biologicals
NBP2-99832-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BirA Polyclonal antibody specifically detects BirA in E. coli samples. It is validated for ELISASpecifications
| BirA | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Epitope Tags | |
| 0.2 um filtered solution in PBS | |
| 948469 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| E. coli | |
| Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase | |
| Produced in rabbits immunized with purified, recombinant E. coli BirA (Uniprot#: P06709; Met1-Lys321). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title