Learn More
MedChemExpress Biotin-VAD-FMK
Specifications
Specifications
| Quantity | 5 mg |
| CAS | 1135688-15-1 |
| Product Type | Biochemical Assay Reagents |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Solubility | DMSO : ≥ 27.3 mg/mL (40.58 mM) |
| Molecular Weight (g/mol) | 672.81 g/mol |