Learn More
Molecular Probes™ Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Brand: Molecular Probes™ T12921
Additional Details : Weight : 0.00450kg
Description
The water-soluble neuronal tracer 5-(and-6)-tetramethylrhodamine biocytin (biocytin TMR) can be detected in fixed tissues by using avidin or streptavidin probes. However, tetramethylrhodamine biocytin can also be observed directly in fixed or unfixed samples because the attached tetramethylrhodamine dye produces bright, orange-fluorescence (absorption/emission maxima ∼554/581 nm).Specifications
Classic Dyes | |
Room Temperature | |
Invitrogen™ |
Store at room temperature and protect from light. | |
Solid | |
Biocytins |
For Research Use Only. Not for use in diagnostic procedures.