Learn More
Agilent BioTek BioCell Vessel and Adapter
For use with: Epoch, Synergy H1, Synergy HTX.
157.00 € - 297.00 €
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
11593535
|
Agilent BioTek
7272051 |
Cuvette | 1cm absorbance measurements |
297.00 €
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12586476
|
Agilent BioTek
7270512 |
Adapter Plate Assembly | 1cm pathlength measurements |
157.00 €
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
BioCell Adapter Plate Assembly can hold up to 8 BioCell cuvets (BT7272051) for 1 cm pathlength measurements in a Agilent BioTek microplate spectrophotometer.Specifications
Epoch, Synergy H1, Synergy HTX |