Learn More
Agilent BioTek BioCell Vessel and Adapter
GREENER_CHOICE
Product Code. 11593535 Shop All Agilent BioTek Products
Click to view available options
Product Type:
Adapter Plate Assembly
Cuvette
For Use With (Application):
1cm absorbance measurements
1cm pathlength measurements
Unit Size:
Each
Product Code. 11593535
Brand: Agilent BioTek 7272051
Please to purchase this item. Need a web account? Register with us today!
For use with: Epoch, Synergy H1, Synergy HTX.
BioCell Adapter Plate Assembly can hold up to 8 BioCell cuvets (BT7272051) for 1 cm pathlength measurements in a Agilent BioTek microplate spectrophotometer.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cuvette
For Use With (Equipment) Epoch, Synergy H1, Synergy HTX
For Use With (Application) 1cm absorbance measurements
Material Quartz