Learn More
Krüss Optronic™ Binocular Microscope
Specifications
Specifications
| Condenser Type | Double lens ABBE Condenser, NA 1.25, with centering, height adjustment and filter holder |
| Eyepiece | 10X/25 mm |
| Head Style | Inclined optical head |
| Lighting Type | Halogen |
| Objective Types | Achromatic: 4X 0.10 NA 17.04 mm working distance, 10X 0.25 NA 8.05 mm working distance, 40X 0.65 NA 0.32 mm working distance, 100X 1.25 NA 0.13 mm working distance |