missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BIK Polyclonal Antibody
GREENER_CHOICE

Product Code. 15904455
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15904455 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15904455 Supplier Invitrogen™ Supplier No. PA578873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Spleen Tissue, Rat Gaster Tissue, Rat Intestine Tissue, MCF-7 whole cell, A549 whole cell, SKOV whole cell. IHC: Mouse Spleen tissue, Rat Spleen tissue, Human Thyroid Cancer tissue. Flow: MCF-7 cell, THP-1 cell, A431 cell.

The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIK
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene BIK
Gene Accession No. O70337, Q13323
Gene Alias apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; BCL2 interacting killer; bcl-2-interacting killer; BCL2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; BIK; Bik-like killer protein; Biklk; BIP 1; BIP1; Blk; BP 4; BP4; cb 60; natural born killer; NBK
Gene Symbols BIK
Host Species Rabbit
Immunogen E.coli-derived human Bik recombinant protein (Position: M1-R123).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114496, 12124, 638
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.