Learn More
Bifenox-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Bifenox-d3 |
| CAS | 2733718-91-5 |
| Molecular Formula | C14 D3 H6 Cl2 N O5 |
| Quantity | 5 mg |
| Synonym | Bifenox D3 (phenyl D3), Bifenox-d3 (2,4-dichlorophenoxy-d3), 2,4-Dichlorophenyl-3,5,6-d3 3-(methoxycarbonyl)-4-nitrophenyl ether, 2,4-Dichlorophenyl-d3 3'-carbomethoxy-4'-nitrophenyl ether, 3-Carbomethoxy-2',4'-dichloro-4-nitrodiphenyl-3',5',6'-d3 ether, Methyl 5-(2,4-dichlorophenoxy-3,5,6-d3)-2-nitrobenzoate |
| InChI Formula | InChI=1S/C14H9Cl2NO5/c1-21-14(18)10-7-9(3-4-12(10)17(19)20)22-13-5-2-8(15)6-11(13)16/h2-7H,1H3/i2D,5D,6D |
| SMILES | [2H]c1c([2H])c(Oc2ccc(c(c2)C(=O)OC)[N+](=O)[O-])c(Cl)c([2H])c1Cl |
| IUPAC Name | methyl 5-(2,4-dichloro-3,5,6-trideuteriophenoxy)-2-nitrobenzoate |
| Molecular Weight (g/mol) | 345.1494 |
| Formula Weight | 344.0046 |