Learn More
B Braun™ Biemer Vessel Clips
Description
- Blade gripping surface with oblique serrations for safe but careful gripping of vessels
- No risk due to rigidly fixed pressure measure which guarantees application without any risk
- Closing force is measured at 1/3 of the blades at 1 mm opening
Specifications
Specifications
| Length (Metric) | 1.4 mm (Jaw) |
| Material | Stainless Steel |
| Gauge | 15 to 20 g |
| Tip Style | 4 mm opening |
| Product Type | Biemer Forceps |