Learn More
Tocris Bioscience™ BI 9564
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
BI 9564 is a potent and selective BRD9 and BRD7 inhibitor (Kd values are 14 and 239 nM, respectively). Exhibits selectivity for BRD9/7 over 48 other bromodomains, 324 kinases and 55 GPCRs. Inhibits proliferation of AML cell lines in vitro and reduces AML tumor xenograft growth in vivo. Cell permeable and orally active.
Specifications
Specifications
| Chemical Name or Material | 4-[4-[(Dimethylamino)methyl]-2,5-dimethoxyphenyl]-2-methyl-1,2-dihydro-2,7-naphthyridin-1-one |
| CAS | 1883429-22-8 |
| Quantity | 10 mg |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C20H23N3O3 |
| Formula Weight | 353.41 |
| Purity | >98% |