missing translation for 'onlineSavingsMsg'
Learn More

BI-847325, MedChemExpress

Product Code. 16453667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453667 50 mg 50mg
16493616 25 mg 25mg
16487556 100 mg 100mg
16437658 10 mg 10mg
16400128 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453667 Supplier Medchem Express Supplier No. HY18955
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BI-847325 is an ATP competitive dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively.

BI-847325 is an ATP competitive dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively.

Specifications

Chemical Name or Material BI-847325
Physical Form Solid
CAS 1207293-36-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H28N4O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 16.67 mg/mL (35.88 mM; Need ultrasonic)
SMILES O=C(NCC)C#CC1=CC(NC/2=O)=C(C=C1)C2=C(NC3=CC=C(CN(C)C)C=C3)/C4=CC=CC=C4
Molecular Weight (g/mol) 464.56
Formula Weight 464.56
Percent Purity 98.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.