missing translation for 'onlineSavingsMsg'
Learn More

BHLHE41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16850544 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16850544 20 μL 20µL
16840544 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16850544 Supplier Proteintech Supplier No. 126881AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.

Specifications

Antigen BHLHE41
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BHLHE41
Gene Accession No. Q99PV5, Q9C0J9
Gene Alias BHLHB3, BHLHE41, DEC2, hDEC2, SHARP 1, SHARP1
Gene Symbols Bhlhe41
Host Species Rabbit
Immunogen BHLHE41 Fusion Protein Ag3381
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117095, 79362, 79365
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.