missing translation for 'onlineSavingsMsg'
Learn More

BGT226 maleate, MedChemExpress

Product Code. p-200101422
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488347

Brand: Medchem Express HY13334

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BGT226 (NVP-BGT226 maleate) is a PI3K (with IC50s of 4 nM, 63 nM and 38 nM for PI3Kα, PI3Kβ and PI3Kγ) /mTOR dual inhibitor which displays potent growth-inhibitory activity against human head and neck cancer cells.

BGT226 (NVP-BGT226 maleate) is a PI3K (with IC50s of 4 nM, 63 nM and 38 nM for PI3Kα, PI3Kβ and PI3Kγ) /mTOR dual inhibitor which displays potent growth-inhibitory activity against human head and neck cancer cells.

Specifications

Chemical Name or Material BGT226 maleate
CAS 1245537-68-1
Color White
Purity Grade Notes Research
Molecular Formula C32H29F3N6O6
Quantity 5 mg
Synonym NVP-BGT226 maleate
Solubility Information DMSO : 100 mg/mL (153.70 mM; Need ultrasonic)
SMILES COC1=CC=C(C=N1)C2=CC=C(C3=C2)N=CC(N4C)=C3N(C4=O)C5=CC=C(C(C(F)(F)F)=C5)N6CCNCC6.OC(/C=C\C(O)=O)=O
Molecular Weight (g/mol) 650.6
Formula Weight 650.6
Percent Purity 98.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.