Learn More
BGT226 maleate, MedChemExpress
Product Code. 16488347
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16488347
Brand: Medchem Express HY13334
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BGT226 (NVP-BGT226 maleate) is a PI3K (with IC50s of 4 nM, 63 nM and 38 nM for PI3Kα, PI3Kβ and PI3Kγ) /mTOR dual inhibitor which displays potent growth-inhibitory activity against human head and neck cancer cells.
BGT226 (NVP-BGT226 maleate) is a PI3K (with IC50s of 4 nM, 63 nM and 38 nM for PI3Kα, PI3Kβ and PI3Kγ) /mTOR dual inhibitor which displays potent growth-inhibitory activity against human head and neck cancer cells.
Specifications
Chemical Name or Material BGT226 maleate
CAS 1245537-68-1
Color White
Purity Grade Notes Research
Molecular Formula C32H29F3N6O6
Quantity 5 mg
Synonym NVP-BGT226 maleate
Solubility Information DMSO : 100 mg/mL (153.70 mM; Need ultrasonic)
SMILES COC1=CC=C(C=N1)C2=CC=C(C3=C2)N=CC(N4C)=C3N(C4=O)C5=CC=C(C(C(F)(F)F)=C5)N6CCNCC6.OC(/C=C\C(O)=O)=O
Molecular Weight (g/mol) 650.6
Formula Weight 650.6
Percent Purity 98.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease