Learn More
Betulinaldehyde, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Betulinaldehyde(Betunal) belongs to pentacyclic triterpenoids and was reported to exhibit antimicrobial activities against bacteria and fungi, including S.
Specifications
Specifications
| Chemical Name or Material | Betulinaldehyde |
| CAS | 13159-28-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H48O2 |
| Quantity | 10 mg |
| Synonym | Betulinic aldehyde Betunal |
| Solubility Information | DMSO : 20 mg/mL (45.38 mM; Need ultrasonic) |
| SMILES | C=C(C)[C@@H]1CC[C@]2(C=O)CC[C@@]3(C)[C@]4(C)CC[C@@]5([H])C(C)(C)[C@@H](O)CC[C@]5(C)[C@@]4([H])CC[C@]3([H])[C@]21[H] |
| Molecular Weight (g/mol) | 440.7 |
| Show More |