missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beta Lactoglobulin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597512
This item is not returnable.
View return policy
Description
Beta Lactoglobulin Polyclonal Antibody for Western Blot, ELISA
Beta-lactoglobulin is a major protein found in dairy products and is not present in human milk. It has been identified as a potential milk allergen, and is responsible for transport of both iron and hydrophobic molecules.
Specifications
| Beta Lactoglobulin | |
| Polyclonal | |
| Unconjugated | |
| PAEP | |
| beta lactoglobulin C; beta lactoglobulin D; beta-lactoglobulin; Beta-LG; BLG; bLG C; Bos d 5; lactoglobulin, beta; LGB; PAEP; progestagen-associated endometrial protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 280838 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P02754 | |
| PAEP | |
| Bovine milk Beta-lactoglobulin (Native Protein). | |
| 100 μg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction