missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Galactosidase-1/GLB1 Antibody (OTI1C9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70829B
This item is not returnable.
View return policy
Description
beta-Galactosidase-1/GLB1 Monoclonal specifically detects beta-Galactosidase-1/GLB1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| beta-Galactosidase-1/GLB1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GLB1 | |
| Full length recombinant protein of human beta-Galactosidase-1/GLB1 (NP_001073279) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1C9 | |
| Western Blot, Immunohistochemistry | |
| Acid beta-galactosidase, beta-galactosidase, EBP, EC 3.2.1.23, Elastin receptor 1, elastin receptor 1 (67kD), elastin receptor 1, 67kDa, ELNR1, galactosidase, beta 1, Lactase, MPS4B | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 2720.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction