missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ beta-Defensin 3 Antibody Blocking Peptide

Product Code. 18528243 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18528243

Brand: Novus Biologicals™ NB200117PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 55894
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB200-117
Gene Alias BD3, BD-3, beta-defensin 103, Beta-defensin 3, DEFB103, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103 B, Defensin-like protein, hBD-3, HBD3HBP3
Gene Symbol DEFB103B
Quantity 0.1 mg
Research Category Biologically Active Proteins, Immunology
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.