missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-D-glucosidase/beta-D-fucosidase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144547
This item is not returnable.
View return policy
Description
beta-D-glucosidase/beta-D-fucosidase Polyclonal Antibody for Western Blot, ELISA
Bifunctional beta-D-glucosidase/beta-D-fucosidase. Activity towards pNP-beta-D-fucoside is about 80-85% of the activity towards pNP-beta-D-glucoside. Also has slight activity (less than 10%) towards pNP-beta-D-galactoside, and very low activity (less than 1%) towards pNP-beta-D-xyloside. Hydrolyzes laminaribiose, sophorose, cellobiose and gentobiose. Not active against maltose, pNP-alpha-D-glucoside or pNP-beta-L-fucoside.
Specifications
| beta-D-glucosidase/beta-D-fucosidase | |
| Polyclonal | |
| Unconjugated | |
| Bifunctional beta-D-glucosidase/beta-D-fucosidase | |
| Recombinant Bifidobacterium breve Bifunctional beta-D-glucosidase/beta-D-fucosidase protein (253-460aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.54 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction