missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Mouse anti-Human, Clone: 13D6, Novus Biologicals™
Mouse Monoclonal Antibody
267.00 € - 598.00 €
Specifications
| Antigen | beta-2 Adrenergic R/ADRB2 |
|---|---|
| Clone | 13D6 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372153
|
Novus Biologicals
NBP3-12007-0.025MG |
0.025 mg |
267.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373312
|
Novus Biologicals
NBP3-12007-0.2MG |
0.2 mg |
598.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunoprecipitationSpecifications
| beta-2 Adrenergic R/ADRB2 | |
| 1 mg/ml | |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS | |
| 154 | |
| IgG1 κ | |
| Protein A purified |
| 13D6 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
| This antibody was raised by immunizing mice with beta-2 adrenergic receptor. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title