missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Antibody (13D6), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12007G
This item is not returnable.
View return policy
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| beta-2 Adrenergic R/ADRB2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 13D6 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
| This antibody was raised by immunizing mice with beta-2 adrenergic receptor. | |
| 0.1 mL | |
| Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
| 154 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction