missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 6797 Chemical Resistant Neoprene Gloves, Extended Sleeve

Product Code. 10301472
Change view
Click to view available options
Size:
10
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10301472 10 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10301472 Supplier SHOWA™ Supplier No. 679710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Smooth surface neoprene glove with cotton liner provides arm-length comfort in hot or cold environments, while protecting against acids and caustics.

  • Extended-arm protection
  • Protection against acids, caustics, oils, greases and many solvents
  • Protection against physical hazards such as abrasion and cut
  • Cotton liner is cool and comfortable in hot conditions and keeps wearer warm in cold conditions
  • Ideal applications include:
    • Chemical spray and treatment
    • Coating preparation
    • Laboratory, pharma and analysis
    • Painting and spray workshops

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 0321, EN 407:3242XX
Applications Laboratory
Color Black
Latex Free Yes
Material Neoprene
Product Type Neoprene Gloves
Size 10
EN388 Standard EN 388:3111X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Smooth
Liner Material Cotton
Lining Lined
Model 6797-10
Series 6797
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.