missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Beryllium Copper, CDA 172, C17200, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Beryllium Copper, CDA 172, C17200, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Chromium/Tin/Manganese/Phosphorus/Lead/Zinc/Nickel/Aluminium/Iron/Silicon/Cobalt/Beryllium/Copper/Nitrogen/Arsenic/Bismuth/Boron/Cadmium/Molybdenum/Oxygen/Selenium/Titanium/Zirconium/Antimony/Magnesium/Silver/Carbon |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Chromium 0.0007 %/Tin 0.0009 %/Manganese 0.0017 %/Phosphorus 0.003 %/Lead 0.005 %/Zinc 0.008 %/Nickel 0.01 %/Aluminium 0.022 %/Iron 0.029 %/Silicon 0.057 %/Cobalt 0.344 %/Beryllium 1.79 %/Copper 97.7 %/Nitrogen (<0.001) %/Arsenic (<0.005) %/Bismuth (<0.005) %/Boron (<0.005) %/Cadmium (<0.005) %/Molybdenum (<0.005) %/Oxygen (<0.005) %/Selenium (<0.005) %/Titanium (<0.005) %/Zirconium (<0.005) %/Antimony (0.001) %/Magnesium (0.0013) %/Silver (0.002) %/Carbon (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction