Learn More
Emma Safety Footwear Berlin Safety Shoes
Product Code. 15978411
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15978411
Brand: Emma Safety Footwear MM40664742
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Woven textile combined with PU provides a wear-resistant polyurethane that optimally protects the textile. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot. Thanks to the Hydro-Tec™ lining, this safety shoe is also particularly breathable.
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile Polyurethane coated construction
  • Metal-free
  • Chrome-free lining
  • Hydro-Tec™ Antibacterial lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Gray, Blue
Design Berlin
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 42
Toe Material Composite