Learn More
Emma Safety Footwear Berlin Safety Shoes
Product Code. 15928421
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15928421
Brand: Emma Safety Footwear MM40664747
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Woven textile combined with PU provides a wear-resistant polyurethane that optimally protects the textile. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot. Thanks to the Hydro-Tec™ lining, this safety shoe is also particularly breathable.
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile Polyurethane coated construction
  • Metal-free
  • Chrome-free lining
  • Hydro-Tec™ Antibacterial lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Gray, Blue
Design Berlin
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 47
Toe Material Composite