Learn More
Emma Safety Footwear Berlin Safety Shoes
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Description
- S1P-Safety shoe with sole plate for dry work
- Woven Textile Polyurethane coated construction
- Metal-free
- Chrome-free lining
- Hydro-Tec™ Antibacterial lining
- ESD certified
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Gray, Blue |
| Design | Berlin |
| Liner Material | Hydro-Tec™ Antibacterial |
| Material | Textile |
| Size | 46 |
| Toe Material | Composite |