missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde dimethyl acetal, 97%

Product Code. 11342636
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342636 50 g 50g
11332636 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342636 Supplier Thermo Scientific Alfa Aesar Supplier No. L08651.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127657-97-0 | C11H16O3 | 196.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127657-97-0
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00082233
InChI Key QITBBLPNYKECAZ-UHFFFAOYSA-N
Synonym benzyloxyacetaldehyde dimethyl acetal, 2,2-dimethoxyethoxy methyl benzene, benzyloxyacetaldehyde dimethylacetal, benzene, 2,2-dimethoxyethoxy methyl, acmc-20alp4, 2,2-dimethoxyethylbenzyl ether, 2-benzyloxy-1,1-dimethoxyethane;, benzyloxy-acetaldehyd-dimethylacetal;
PubChem CID 7021458
IUPAC Name 2,2-dimethoxyethoxymethylbenzene
SMILES COC(COCC1=CC=CC=C1)OC

Specifications

Density 1.035
Boiling Point 80°C to 82°C (0.5 mmHg)
Refractive Index 1.491
Quantity 50 g
Beilstein 3261948
Formula Weight 196.24
Percent Purity 97%
Chemical Name or Material Benzyloxyacetaldehyde dimethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.