Learn More
(Benzyloxy)(methyl)amine, TRC
Specifications
Specifications
| Chemical Name or Material | (Benzyloxy)(methyl)amine |
| CAS | 22513-22-0 |
| Molecular Formula | C8H11NO |
| Quantity | 1 g |
| Synonym | N-(Phenylmethoxy)methanamine, (Benzyloxy)(methyl)amine, N-Benzyloxy-N-methylamine, N-Methyl-O-benzylhydroxylamine, O-Benzyl-N-methylhydroxylamine |
| InChI Formula | InChI=1S/C8H11NO/c1-9-10-7-8-5-3-2-4-6-8/h2-6,9H,7H2,1H3 |
| SMILES | CNOCc1ccccc1 |
| IUPAC Name | N-phenylmethoxymethanamine |
| Molecular Weight (g/mol) | 137.17 |
| Formula Weight | 137.0841 |