missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyl-n-tetradecylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Product Code. 15450307
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450307 5 g 5g
15460307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450307 Supplier Thermo Scientific Alfa Aesar Supplier No. J63465.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Phase transfer catalyst

Phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-08-2
Molecular Formula C23H42ClN
Molecular Weight (g/mol) 368.05
MDL Number MFCD00011771
InChI Key OCBHHZMJRVXXQK-UHFFFAOYSA-M
Synonym miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
PubChem CID 8755
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Color White
Linear Formula CH3(CH2)13N(CH3)2(CH2C6H5)Cl
Quantity 5 g
UN Number UN3261
Beilstein 4062599
Merck Index 14,1059
IUPAC Name benzyldimethyltetradecylazanium chloride
Formula Weight 368.05 (Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzyldimethyl-n-tetradecylammonium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.