Learn More
Benzyl Violet (Technical Grade), TRC
Product Code. 17952755
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952755 250 mg 250mg
17904335 500 mg 500mg
17931645 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17952755 Supplier Toronto Research Chemicals Supplier No. TRCB316553250MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1694-09-3
Molecular Formula C39 H40 N3 O6 S2 . Na
Molecular Weight (g/mol) 733.87
Synonym Acid Violet 5B (6CI), Benzenemethanaminium, N-[4-[[4-(dimethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, hydroxide, inner salt, sodium salt, Benzenemethanaminium, N-[4-[[4-(dimethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (9CI), C.I. Acid Violet 49 (7CI), C.I. Acid Violet 49, sodium salt (8CI), 11386 Violet, A.F. Violet No. 1, Acid Fast Violet 5BN, Acid Milling Violet 4BS, Acid Violet, Acid Violet 23184, Acid Violet 49, Acid Violet 4BNP, Acid Violet 5BN, Acid Violet 6B, Acid Violet S, Acilan Violet S 4BN, Aizen Acid Violet 5BH, Atlantic Acid Violet 4BNS, Benzyl Violet, Benzyl Violet 3B, Benzyl Violet 4B, Borunil Violet A-6B, C.I. 42640, C.I. Food Violet 2, Cogilor Violet 411.12, Coomassie Violet, D And C Violet No. 1, Dinacid Violet 4B, Dycosacid Violet 5B, Everacid Violet 4BR, FD and C Violet 1, FD and C Violet No. 1, FD&C Violet No. 1, Fast Acid Violet 5BN, Food Violet 2, Hidacid Wool Violet 5B, Ichoacid Violet M, Indacid Violet S 4BN, Kenanthrol Violet 4BNS, Kiton Violet 4BNS, Libacid Violet L4BS, Lowalan Violet 49, Monacid Violet 4BS, Old Japan Purple 1, Old Japan Violet 1, Orbacid Violet S 4B, Orient Water Violet 1, Pacid Violet 4BS, Pergacid Violet 2B, Ratna Acid Violet 5BND, Ravi Acid Violet 4B, Serva Violet 49, Tertracid Brilliant Violet 6B, Triacid Violet 4BN, Violet 6B, Violet No. 1, Water Violet 1, Wool Violet 4BN, Wool Violet 5BN
IUPAC Name sodium;3-[[4-[[4-(dimethylamino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMILES [Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccc(cc5)N(C)C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C39H41N3O6S2.Na/c1-5-41(27-29-9-7-11-37(25-29)49(43,44)45)35-21-15-32(16-22-35)39(31-13-19-34(20-14-31)40(3)4)33-17-23-36(24-18-33)42(6-2)28-30-10-8-12-38(26-30)50(46,47)48;/h7-26H,5-6,27-28H2,1-4H3,(H-,43,44,45,46,47,48);/q;+1/p-1
Formula Weight 733.2256
Chemical Name or Material Acid Violet 49