missing translation for 'onlineSavingsMsg'
Learn More

Benzyl thiocyanate, 98+%, Thermo Scientific™

Product Code. 11456756
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456756 250 g 250g
11446756 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456756 Supplier Thermo Scientific Alfa Aesar Supplier No. B23902.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-37-1
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00001832
InChI Key ABNDFSOIUFLJAH-UHFFFAOYSA-N
Synonym tropeolin, thiocyanic acid, phenylmethyl ester, solvat 14, benzylthiocyanate, benzylrhodonid, thiocyanic acid, benzyl ester, alpha-thiocyanatotoluene, benzyl-thiocyanate, thiocyanic acid benzyl ester, phenylmethyl thiocyanate
PubChem CID 18170
ChEBI CHEBI:16017
IUPAC Name benzyl thiocyanate
SMILES N#CSCC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Density 1.32
Boiling Point 230°C
Flash Point 110°C (230°F)
Odor Irritating
Quantity 250 g
UN Number UN2811
Beilstein 1859726
Sensitivity Moisture sensitive
Formula Weight 149.22
Percent Purity ≥98%
Chemical Name or Material Benzyl thiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.