Learn More
Benzyl Salicylate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl Salicylate-d4 |
| CAS | 1219802-40-0 |
| Molecular Formula | C14 D4 H8 O3 |
| Quantity | 25 mg |
| Synonym | 6-Hydroxybenzoic-2,3,4,5-d4 acid phenylmethyl ester, 2-Hydroxybenzoic-3,4,5,6-d4 acid benzyl ester, Salicylic-d4 acid benzyl ester, Salicylic Acid Benzyl Ester-d4, Salicylic Acid Benzyl Ester-D4 (salicyl-D4), Benzyl Salicylate-d4, Benzyl 2-Hydroxybenzoate-3,4,5,6-d4 |
| InChI Formula | InChI=1S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15H,10H2/i4D,5D,8D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCc2ccccc2)c(O)c1[2H] |
| IUPAC Name | benzyl 2,3,4,5-tetradeuterio-6-hydroxybenzoate |
| Molecular Weight (g/mol) | 232.2679 |
| Formula Weight | 232.1038 |