missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Isothiocyanate, TRC

Product Code. 30005009
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005009 5 g 5g
30134928 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005009 Supplier Toronto Research Chemicals Supplier No. TRCB2815005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-78-6
Formula Weight 149.0299
InChI Formula InChI=1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2
IUPAC Name isothiocyanatomethylbenzene
Molecular Formula C8 H7 N S
Molecular Weight (g/mol) 149.21
SMILES S=C=NCc1ccccc1
Synonym Isothiocyanic Acid Benzyl Ester, (Isothiocyanatomethyl)benzene, 1-(Isothiocyanatomethyl)benzene, AB 2, Benzyl Mustard Oil, NSC 118976, Phenylmethyl Isothiocyanate, Tromacaps, Tromalyt, Urogran;
Chemical Name or Material Benzyl Isothiocyanate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.