Learn More
Benzyl-8-azabicyclo[3.2.1]octan-3-exo-amine, TRC
Product Code. 30341129
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341129 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30341129 Supplier Toronto Research Chemicals Supplier No. TRCB230760500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzyl-8-azabicyclo[3.2.1]octan-3-exo-amine
CAS 76272-36-1
Molecular Formula C14 H20 N2
Quantity 500 mg
InChI Formula InChI=1S/C14H20N2/c15-12-8-13-6-7-14(9-12)16(13)10-11-4-2-1-3-5-11/h1-5,12-14H,6-10,15H2/t12-,13-,14+
SMILES N[C@@H]1C[C@H]2CC[C@@H](C1)N2Cc3ccccc3
IUPAC Name (1R,5S)-8-benzyl-8-azabicyclo[3.2.1]octan-3-amine
Molecular Weight (g/mol) 216.32
Formula Weight 216.1626