Learn More
Benzyl (4-Hydroxyphenyl)acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl (4-Hydroxyphenyl)acetate |
| CAS | 27727-37-3 |
| Molecular Formula | C15H14O3 |
| Quantity | 250 mg |
| Synonym | 4-Hydroxy-benzeneacetic Acid Phenylmethyl Ester, (p-Hydroxyphenyl)acetic Acid Benzyl Ester |
| InChI Formula | InChI=1S/C15H14O3/c16-14-8-6-12(7-9-14)10-15(17)18-11-13-4-2-1-3-5-13/h1-9,16H,10-11H2 |
| SMILES | Oc1ccc(CC(=O)OCc2ccccc2)cc1 |
| IUPAC Name | benzyl 2-(4-hydroxyphenyl)acetate |
| Molecular Weight (g/mol) | 242.27 |
| Formula Weight | 242.0943 |