Learn More
Benzyl ((3aS,4R,6S,6aR)-6-(2-Hydroxyethoxy)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl ((3aS,4R,6S,6aR)-6-(2-Hydroxyethoxy)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)carbamate |
| CAS | 274693-54-8 |
| Molecular Formula | C18H25NO6 |
| Quantity | 250 mg |
| Synonym | Ticagrelor Impurity (Intermediate) Tgk-10 |
| InChI Formula | InChI=1S/C18H25NO6/c1-18(2)24-15-13(10-14(16(15)25-18)22-9-8-20)19-17(21)23-11-12-6-4-3-5-7-12/h3-7,13-16,20H,8-11H2,1-2H3,(H,19,21)/t13-,14+,15+,16-/m1/s1 |
| SMILES | O=C(OCC1=CC=CC=C1)N[C@H]2[C@@]3([H])[C@@](OC(C)(C)O3)([H])[C@@H](OCCO)C2 |
| IUPAC Name | benzyl N-[(3aR,4S,6R,6aS)-4-(2-hydroxyethoxy)-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]carbamate |
| Molecular Weight (g/mol) | 351.39 |
| Formula Weight | 351.1682 |