Learn More
Benzyl 3-(4-Hydroxyphenyl)propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl 3-(4-Hydroxyphenyl)propionate |
| CAS | 31770-76-0 |
| Molecular Formula | C16 H16 O3 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C16H16O3/c17-15-9-6-13(7-10-15)8-11-16(18)19-12-14-4-2-1-3-5-14/h1-7,9-10,17H,8,11-12H2 |
| SMILES | Oc1ccc(CCC(=O)OCc2ccccc2)cc1 |
| IUPAC Name | benzyl 3-(4-hydroxyphenyl)propanoate |
| Molecular Weight (g/mol) | 256.3 |
| Formula Weight | 256.1099 |