Learn More
Benzyl 2,3,4-Tri-O-benzyl-Beta-D-glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl 2,3,4-Tri-O-benzyl-beta-D-glucopyranoside |
| CAS | 27851-29-2 |
| Molecular Formula | C34 H36 O6 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C34H36O6/c35-21-30-31(36-22-26-13-5-1-6-14-26)32(37-23-27-15-7-2-8-16-27)33(38-24-28-17-9-3-10-18-28)34(40-30)39-25-29-19-11-4-12-20-29/h1-20,30-35H,21-25H2/t30-,31-,32+,33-,34-/m1/s1 |
| SMILES | OC[C@H]1O[C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@@H](OCc4ccccc4)[C@@H]1OCc5ccccc5 |
| IUPAC Name | [(2R,3R,4S,5R,6R)-3,4,5,6-tetrakis(phenylmethoxy)oxan-2-yl]methanol |
| Molecular Weight (g/mol) | 540.65 |
| Formula Weight | 540.2512 |