Learn More
Benzoylaconine, MedChemExpress
Product Code. 16461338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16461338
Brand: Medchem Express HYN0217
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Benzoylaconine(Isaconitine; Pikraconitin) is an alkaloid in the Chinese traditional medicine Radix Aconiti Lateralis Preparata (Fuzi).
Benzoylaconine(Isaconitine
Specifications
Chemical Name or Material Benzoylaconine
CAS 466-24-0
Color White
Purity Grade Notes Research
Molecular Formula C32H45NO10
Quantity 5 mg
Synonym Isaconitine Pikraconitin
Solubility Information DMSO : 55 mg/mL (91.10 mM; Need ultrasonic)
SMILES CO[C@@H]1C(C(N(CC)C2)C3[C@@H]4OC)([C@@](C[C@@]5(O)[C@@H]6OC(C7=CC=CC=C7)=O)([H])[C@@]6([H])[C@@]3([C@@H](O)[C@@H]5OC)O)[C@@]4([H])[C@@]2(COC)[C@H](O)C1
Molecular Weight (g/mol) 603.7
Formula Weight 603.7
Percent Purity 98.1%
Grade Research
Physical Form Powder