Learn More
Benzophenone-4-carboxamidocysteine Methanethiosulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzophenone-4-carboxamidocysteine Methanethiosulfonate |
| CAS | 317821-69-5 |
| Molecular Formula | C18 H17 N O6 S2 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C18H17NO6S2/c1-27(24,25)26-11-15(18(22)23)19-17(21)14-9-7-13(8-10-14)16(20)12-5-3-2-4-6-12/h2-10,15H,11H2,1H3,(H,19,21)(H,22,23)/t15-/m0/s1 |
| SMILES | CS(=O)(=O)SC[C@H](NC(=O)c1ccc(cc1)C(=O)c2ccccc2)C(=O)O |
| IUPAC Name | (2R)-2-[(4-benzoylbenzoyl)amino]-3-methylsulfonylsulfanylpropanoic acid |
| Molecular Weight (g/mol) | 407.46 |
| Formula Weight | 407.0497 |