Learn More
Benzophenone-13C1 (carbonyl-13C), CDN
Specifications
Specifications
| Chemical Name or Material | Benzophenone-13 C |
| CAS | 32488-48-5 |
| Molecular Formula | 13 C C12 H10 O |
| Quantity | 0.5 g |
| Synonym | Methanone-13 C, diphenyl- (9 CI, ACI), Diphenylmethanone-13 C (ACI), Benzophenone-carbonyl-13 C, Carbonyl-13 C-benzophenone, Benzophenone-13C1 (carbonyl-13 C), Benzophenone-13 C (carbonyl-13 C) |
| InChI Formula | InChI=1 S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10 H/i13+1 |
| SMILES | O=[13 C](c1ccccc1)c2ccccc2 |
| IUPAC Name | diphenyl(1 ^{13}C)methanone |
| Molecular Weight (g/mol) | 183.21 |
| Formula Weight | 183.0765 g/mol |
| Show More |